2-Acetoxycinnamic acid ≥95.0% (by GC, titration analysis)

Supplier: TCI America

A2413-1G A2413-5G
TCA2413-1GEA 50.56 CAD
TCA2413-1G TCA2413-5G
2-Acetoxycinnamic acid ≥95.0% (by GC, titration analysis)
2-Acetoxycinnamic acid

CAS Number: 55620-18-3
MDL Number: MFCD00014375
Molecular Formula: C11H10O4
Molecular Weight: 206.20
Purity/Analysis Method: >95.0% (GC,T)
Form: Crystal
Melting point (°C): 156


Product Categories: Bioscience, Phenylpropanoids
Other Product Names / Synonyms: 3-(2-Acetoxyphenyl)-2-propenoic Acid

Formula: C₁₁H₁₀O₄
Melting Pt: 156 °C
MDL Number: MFCD00014375
CAS Number: 55620-18-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=06b667fe9b70f250d6e378b8642edd54feb791cd&vid=06b667fe9b70f250d6e378b8642edd54feb791cd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR