N-(6-Aminohexyl)-1-naphthalenesulfonamide hydrochloride ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: W-5 Hydrochloride

A2410-25MG A2410-500MG
TCA2410-25MGEA 41.08 CAD
TCA2410-25MG TCA2410-500MG
N-(6-Aminohexyl)-1-naphthalenesulfonamide hydrochloride ≥98.0% (by HPLC, total nitrogen)
N-(6-Aminohexyl)-1-naphthalenesulfonamide hydrochloride

CAS Number: 61714-25-8
MDL Number: MFCD00058043
Molecular Formula: C16H22N2O2S
Molecular Weight: 342.88
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 164
Storage Temperature: <0°C


Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Calmodulin Antagonists
Other Product Names / Synonyms: W-5 Hydrochloride

Formula: C₁₆H₂₂N₂O₂S
MW: 342.88 g/mol
Melting Pt: 164 °C
MDL Number: MFCD00058043
CAS Number: 61714-25-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f9cece272158f06548f16383b8fcc93e12f039cd&vid=f9cece272158f06548f16383b8fcc93e12f039cd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR