N-(6-Aminohexyl)-5-chloro-1-naphthalenesulfonamide hydrochloride ≥98.0%

Supplier: TCI America

Synonyms: W-7 Hydrochloride, N-(6-Aminohexyl)-5-chloronaphthalene-1-sulfonamide hydrochloride, N-(6-Aminohexyl)-5-chloronaphthalene-1-sulphonamide hydrochloride

A2409-100MG A2409-1G
TCA2409-100MGEA 289.3 CAD
TCA2409-100MG TCA2409-1G
N-(6-Aminohexyl)-5-chloro-1-naphthalenesulfonamide hydrochloride ≥98.0%
N-(6-Aminohexyl)-5-chloro-1-naphthalenesulfonamide hydrochloride

CAS Number: 61714-27-0
MDL Number: MFCD00012559
Molecular Formula: C16H21ClN2O2S
Molecular Weight: 377.32
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 222
Storage Temperature: <0°C


Product Categories: Bioscience, Calmodulin Antagonists
Other Product Names / Synonyms: W-7 Hydrochloride

Formula: C₁₆H₂₁ClN₂O₂S
MW: 377.32 g/mol
Melting Pt: 222 °C
MDL Number: MFCD00012559
CAS Number: 61714-27-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=757c6ad1fdaea72ee2b273d3b7584c817d9cc426&vid=757c6ad1fdaea72ee2b273d3b7584c817d9cc426&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR