N-(4-Aminobutyl)-2-naphthalenesulfonamide hydrochloride ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: W-12 Hydrochloride

A2408-25MG A2408-500MG
TCA2408-25MGEA 34.76 CAD
TCA2408-25MG TCA2408-500MG
N-(4-Aminobutyl)-2-naphthalenesulfonamide hydrochloride ≥98.0% (by HPLC, total nitrogen)
N-(4-Aminobutyl)-2-naphthalenesulfonamide hydrochloride

CAS Number: 89108-46-3
MDL Number: MFCD00058052
Molecular Formula: C14H18N2O2S
Molecular Weight: 314.83
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 183
Storage Temperature: 0-10°C


Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Calmodulin Antagonists
Other Product Names / Synonyms: W-12 Hydrochloride

Formula: C₁₄H₁₈N₂O₂S
MW: 314.83 g/mol
Melting Pt: 183 °C
MDL Number: MFCD00058052
CAS Number: 89108-46-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9b544ae6f4a7f130d436e26989788361aa2a63bc&vid=9b544ae6f4a7f130d436e26989788361aa2a63bc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR