3-Amino-2-chloro-5-methylpyridine ≥98.0%

Supplier: TCI America

Synonyms: 2-Chloro-5-methylpyridin-3-amine

A2405-25G A2405-5G
TCA2405-25GEA 319.16 CAD
TCA2405-25G TCA2405-5G
3-Amino-2-chloro-5-methylpyridine ≥98.0%
3-Amino-2-chloro-5-methylpyridine

CAS Number: 34552-13-1
MDL Number: MFCD03427656
Molecular Formula: C6H7ClN2
Molecular Weight: 142.59
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 89


Other Product Names / Synonyms: 5-Amino-6-chloro-3-picoline

Formula: C₆H₇ClN₂
MW: 142.5900 g/mol
Melting Pt: 89 °C
MDL Number: MFCD03427656
CAS Number: 34552-13-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=192298bb6f52c2d74d04ba56667926ab534dd923&vid=192298bb6f52c2d74d04ba56667926ab534dd923&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR