2-Acetyl-6-bromopyridine ≥98.0%

Supplier: TCI America

A2404-1G A2404-5G
TCA2404-1GEA 66.36 CAD
TCA2404-1G TCA2404-5G
2-Acetyl-6-bromopyridine ≥98.0%
2-Acetyl-6-bromopyridine

CAS Number: 49669-13-8
MDL Number: MFCD00272200
Molecular Formula: C7H6BrNO
Molecular Weight: 200.04
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 53

Formula: C₇H₆BrNO
MW: 200.03 g/mol
Melting Pt: 53 °C
MDL Number: MFCD00272200
CAS Number: 49669-13-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=28032f821add74c7ebbad085818b393c0ce6411d&vid=28032f821add74c7ebbad085818b393c0ce6411d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR