3-Amino-2-methoxypyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2-Methoxypyridin-3-amine, 2-Methoxy-3-pyridylamine

A2403-25G A2403-5G
TCA2403-25GEA 338.12 CAD
TCA2403-25G TCA2403-5G
3-Amino-2-methoxypyridine ≥98.0% (by GC, titration analysis)
3-Amino-2-methoxypyridine

CAS Number: 20265-38-7
MDL Number: MFCD00833386
Molecular Formula: C6H8N2O
Molecular Weight: 124.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 118
Melting point (°C): 67

Formula: C₆H₈N₂O
MW: 124.14 g/mol
Boiling Pt: 118 °C
Melting Pt: 67 °C
MDL Number: MFCD00833386
CAS Number: 20265-38-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d5c6a5b0c5cad018baab6899756738b7b8d9086f&vid=d5c6a5b0c5cad018baab6899756738b7b8d9086f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR