2-Amino-3-bromo-5-chloropyridine ≥98.0%

Supplier: TCI America

Synonyms: 3-Bromo-5-chloro-2-pyridinamine, 3-Bromo-5-chloro-2-pyridylamine, 3-Bromo-5-chloropyridin-2-amine

A2402-25G A2402-5G
TCA2402-25GEA 213.3 CAD
TCA2402-25G TCA2402-5G
2-Amino-3-bromo-5-chloropyridine ≥98.0%
2-Amino-3-bromo-5-chloropyridine

CAS Number: 26163-03-1
MDL Number: MFCD00955627
Molecular Formula: C5H4BrClN2
Molecular Weight: 207.46
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 84

Formula: C₅H₄BrClN₂
MW: 207.46
Melting Pt: 84 °C
MDL Number: MFCD00955627
CAS Number: 26163-03-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9c9a324c67736c3abfc7f1294a898bedced02a4d&vid=9c9a324c67736c3abfc7f1294a898bedced02a4d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR