2-Amino-4-fluorophenol ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 5-Fluoro-2-hydroxyaniline

A2400-25G A2400-5G
TCA2400-25GEA 2679.68 CAD
TCA2400-25G TCA2400-5G
2-Amino-4-fluorophenol ≥97.0% (by titrimetric analysis)
2-Amino-4-fluorophenol

CAS Number: 399-97-3
MDL Number: MFCD00077451
Molecular Formula: C6H6FNO
Molecular Weight: 127.12
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 146


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 5-Fluoro-2-hydroxyaniline

Formula: C₆H₆FNO
MW: 127.12 g/mol
Melting Pt: 146 °C
MDL Number: MFCD00077451
CAS Number: 399-97-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3e9cd73ea147761c467656cf59f4694ff1e89d29&vid=3e9cd73ea147761c467656cf59f4694ff1e89d29&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR