5-Chloro-3-methylanthranilic acid ≥98.0%

Supplier: TCI America

Synonyms: 2-Amino-5-chloro-3-methylbenzoic acid

A2399-25G A2399-5G
TCA2399-25GEA 154.84 CAD
TCA2399-25G TCA2399-5G
5-Chloro-3-methylanthranilic acid ≥98.0%
5-Chloro-3-methylanthranilic acid

CAS Number: 20776-67-4
MDL Number: MFCD02358895
Molecular Formula: C8H8ClNO2
Molecular Weight: 185.61
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 238


Other Product Names / Synonyms: 5-Chloro-3-methylanthranilic Acid, 2-Amino-5-chloro-m-toluic Acid

Formula: C₈H₈ClNO₂
MW: 185.6076 g/mol
Melting Pt: 238 °C
MDL Number: MFCD02358895
CAS Number: 20776-67-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=243f3b3aa565c23458a42855e2aa9e35cf6eef19&vid=243f3b3aa565c23458a42855e2aa9e35cf6eef19&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR