3-Amino-4-chloropyridine ≥98.0%

Supplier: TCI America

Synonyms: 4-Chloro-3-pyridylamine, 4-Chloropyridin-3-amine

A2395-1G A2395-5G
TCA2395-1GEA 67.94 CAD
TCA2395-1G TCA2395-5G
3-Amino-4-chloropyridine ≥98.0%
3-Amino-4-chloropyridine

CAS Number: 20511-15-3
MDL Number: MFCD00235124
Molecular Formula: C5H5ClN2
Molecular Weight: 128.56
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 60
Storage Temperature: 0-10°C

Formula: C₅H₅ClN₂
MW: 128.56 g/mol
Melting Pt: 60 °C
MDL Number: MFCD00235124
CAS Number: 20511-15-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=02a25ac5748e9c500ac055f0c415439c8a18fa89&vid=02a25ac5748e9c500ac055f0c415439c8a18fa89&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR