Aniracetam ≥98.0% (by GC)

Supplier: TCI America

A2394-1G A2394-5G
TCA2394-1GEA 115.34 CAD
TCA2394-1G TCA2394-5G
Aniracetam ≥98.0% (by GC)
Aniracetam

CAS Number: 72432-10-1
MDL Number: MFCD00153767
Molecular Formula: C12H13NO3
Molecular Weight: 219.24
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 122


Product Categories: Bioscience, Other Pharmaceutical Ingredients
Other Product Names / Synonyms: 1-(4-Methoxybenzoyl)-2-pyrrolidone

Formula: C₁₂H₁₃NO₃
Melting Pt: 122 °C
MDL Number: MFCD00153767
CAS Number: 72432-10-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3b6e5a5f530e545a3e19f0c10d79dfcb7d01a302&vid=3b6e5a5f530e545a3e19f0c10d79dfcb7d01a302&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR