Atropic acid ≥93.0% (by GC)
Supplier: TCI America
A2390-25G
A2390-5G
TCA2390-25GEA
300.2
CAD
TCA2390-25G
TCA2390-5G
Atropic acid ≥93.0% (by GC)
Atropic acid
CAS Number: 492-38-6
MDL Number: MFCD00046531
Molecular Formula: C9H8O2
Molecular Weight: 148.16
Purity/Analysis Method: >93.0% (GC)
Form: Crystal
Boiling point (°C): 267
Melting point (°C): 107
Other Product Names / Synonyms: 2-Phenylacrylic Acid
Formula:
C₉H₈O₂ Boiling Pt: 267 °C Melting Pt: 107 °C |
MDL Number:
MFCD00046531 CAS Number: 492-38-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...