5-Aminopicolinic acid ≥98.0% (by HPLC)
Supplier: TCI America
A2386-1G
A2386-5G
TCA2386-1GEA
97.96
CAD
TCA2386-1G
TCA2386-5G
5-Aminopicolinic acid ≥98.0% (by HPLC)
5-Aminopicolinic acid
CAS Number: 24242-20-4
MDL Number: MFCD02684600
Molecular Formula: C6H6N2O2
Molecular Weight: 138.13
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 218
Other Product Names / Synonyms: 5-Aminopicolinic Acid
Formula:
C₆H₆N₂O₂ Melting Pt: 218 °C |
MDL Number:
MFCD02684600 CAS Number: 24242-20-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...