4-Amino-3-fluoropyridine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
A2383-1G
A2383-5G
TCA2383-1GEA
131.79
CAD
TCA2383-1G
TCA2383-5G
4-Amino-3-fluoropyridine ≥98.0% (by GC, titration analysis)
4-Amino-3-fluoropyridine
CAS Number: 2247-88-3
MDL Number: MFCD00234072
Molecular Formula: C5H5FN2
Molecular Weight: 112.11
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 79
Formula:
C₅H₅FN₂ Melting Pt: 79 °C |
MDL Number:
MFCD00234072 CAS Number: 2247-88-3 UN: 2811 ADR: 6.1,II |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...