2-Azidoethyl-2,3,4,6-tetra-O-acetyl-β-D-glucopyranoside ≥92.0% (by HPLC)
Supplier: TCI America
A2377-1G
A2377-5G
TCA2377-1GEA
482.69
CAD
TCA2377-1G
TCA2377-5G
2-Azidoethyl-2,3,4,6-tetra-O-acetyl-β-D-glucopyranoside ≥92.0% (by HPLC)
2-Azidoethyl-2,3,4,6-tetra-O-acetyl-beta-D-glucopyranoside
CAS Number: 140428-81-5
Molecular Formula: C16H23N3O10
Molecular Weight: 417.37
Purity/Analysis Method: >92.0% (HPLC)
Form: Crystal
Melting point (°C): 115
Specific rotation [a]20/D: -35 deg (C=1.5, CHCl3)
Storage Temperature: 0-10°C
Formula:
C₁₆H₂₃N₃O₁₀ Melting Pt: 115 °C |
CAS Number:
140428-81-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...