3-Acetyl-5-bromopyridine ≥97.0%
Supplier: TCI America
A2376-1G
A2376-5G
TCA2376-1GEA
52.93
CAD
TCA2376-1G
TCA2376-5G
3-Acetyl-5-bromopyridine ≥97.0%
3-Acetyl-5-bromopyridine
CAS Number: 38940-62-4
MDL Number: MFCD03086033
Molecular Formula: C7H6BrNO
Molecular Weight: 200.04
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 92
Formula:
C₇H₆BrNO MW: 200.03 g/mol Melting Pt: 92 °C |
MDL Number:
MFCD03086033 CAS Number: 38940-62-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...