2-Acetamido-5-bromopyridine ≥98.0% (by GC)
Supplier: TCI America
A2375-1G
A2375-5G
TCA2375-1GEA
26.86
CAD
TCA2375-1G
TCA2375-5G
2-Acetamido-5-bromopyridine ≥98.0% (by GC)
2-Acetamido-5-bromopyridine
CAS Number: 7169-97-3
MDL Number: MFCD00468968
Molecular Formula: C7H7BrN2O
Molecular Weight: 215.05
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 176
Formula:
C₇H₇BrN₂O Melting Pt: 176 °C |
MDL Number:
MFCD00468968 CAS Number: 7169-97-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...