5-Amino-2-methylbenzoic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

A2373-25G A2373-5G
TCA2373-25GEA 496.12 CAD
TCA2373-25G TCA2373-5G
5-Amino-2-methylbenzoic acid ≥98.0% (by titrimetric analysis)
5-Amino-2-methylbenzoic acid

CAS Number: 2840-04-2
MDL Number: MFCD06208351
Molecular Formula: C8H9NO2
Molecular Weight: 151.17
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 197
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 5-Amino-o-toluic Acid

Formula: C₈H₉NO₂
Melting Pt: 197 °C
MDL Number: MFCD06208351
CAS Number: 2840-04-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=89021897cdcac5ea1c46f635435f59d5ddaf2b30&vid=89021897cdcac5ea1c46f635435f59d5ddaf2b30&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR