2-Amino-6-methoxy-3-nitropyridine ≥97.0% (by GC)
Supplier: TCI America
A2372-25G
A2372-5G
TCA2372-25GEA
679.4
CAD
TCA2372-25G
TCA2372-5G
2-Amino-6-methoxy-3-nitropyridine ≥97.0% (by GC)
2-Amino-6-methoxy-3-nitropyridine
CAS Number: 73896-36-3
MDL Number: MFCD03206481
Molecular Formula: C6H7N3O3
Molecular Weight: 169.14
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 171
Formula:
C₆H₇N₃O₃ Melting Pt: 171 °C |
MDL Number:
MFCD03206481 CAS Number: 73896-36-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...