4-Amino-2,6-dichloropyridine ≥97.0% (by GC, total nitrogen)

Supplier: TCI America

A2369-1G A2369-5G
TCA2369-1GEA 64.78 CAD
TCA2369-1G TCA2369-5G
4-Amino-2,6-dichloropyridine ≥97.0% (by GC, total nitrogen)
4-Amino-2,6-dichloropyridine

CAS Number: 2587-02-2
MDL Number: MFCD00052832
Molecular Formula: C5H4Cl2N2
Molecular Weight: 163.00
Purity/Analysis Method: >97.0% (GC,N)
Form: Crystal
Melting point (°C): 175

Formula: C₅H₄Cl₂N₂
Melting Pt: 175 °C
MDL Number: MFCD00052832
CAS Number: 2587-02-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=69577c0ca1f737cef9011840855b4cef70441e71&vid=69577c0ca1f737cef9011840855b4cef70441e71&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR