3-Amino-4-methylbenzamide ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

A2367-25G A2367-5G
TCA2367-25GEA 222.78 CAD
TCA2367-25G TCA2367-5G
3-Amino-4-methylbenzamide ≥98.0% (by HPLC, titration analysis)
3-Amino-4-methylbenzamide

CAS Number: 19406-86-1
MDL Number: MFCD00035936
Molecular Formula: C8H10N2O
Molecular Weight: 150.18
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 131


Other Product Names / Synonyms: 3-Amino-p-toluamide

Formula: C₈H₁₀N₂O
Melting Pt: 131 °C
MDL Number: MFCD00035936
CAS Number: 19406-86-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=704a8c795ec8810f86ec282bdfd50cfffe91dd08&vid=704a8c795ec8810f86ec282bdfd50cfffe91dd08&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR