(S)-2-Acetamido-3-methylbutanoic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
A2366-25G
A2366-5G
TCA2366-25GEA
74.26
CAD
TCA2366-25G
TCA2366-5G
(S)-2-Acetamido-3-methylbutanoic acid ≥98.0% (by HPLC, titration analysis)
(S)-2-Acetamido-3-methylbutanoic acid
CAS Number: 96-81-1
MDL Number: MFCD00066066
Molecular Formula: C7H13NO3
Molecular Weight: 159.19
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 165
Specific rotation [a]20/D: 13 deg (C=1, AcOH)
Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: Ac-Val-OH
Formula:
C7H13NO3 Melting Pt: 165 °C |
MDL Number:
MFCD00066066 CAS Number: 96-81-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...