O-Acetyl-N-methylhydroxylamine hydrochloride ≥98.0%

Supplier: TCI America

A2365-1G A2365-5G
TCA2365-1GEA 203.35 CAD
TCA2365-1G TCA2365-5G
O-Acetyl-N-methylhydroxylamine hydrochloride ≥98.0%
O-Acetyl-N-methylhydroxylamine hydrochloride

CAS Number: 19689-95-3
Molecular Formula: C3H7NO2
Molecular Weight: 125.55
Purity/Analysis Method: >98.0% (N,T)
Form: Crystal
Melting point (°C): 111
Storage Temperature: 0-10°C

Formula: C₃H₇NO₂
MW: 125.55 g/mol
Melting Pt: 111 °C
CAS Number: 19689-95-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0bd57bd2d41cc208711238db1c4cc5204ac2cef8&vid=0bd57bd2d41cc208711238db1c4cc5204ac2cef8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR