5-Acetylthiophene-2-boronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

A2364-25G A2364-5G
TCA2364-25GEA 1000.14 CAD
TCA2364-25G TCA2364-5G
5-Acetylthiophene-2-boronic acid (contains varying amounts of Anhydride)
5-Acetylthiophene-2-boronic acid

CAS Number: 206551-43-1
MDL Number: MFCD01075681
Molecular Formula: C6H7BO3S
Molecular Weight: 169.99
Form: Crystal
Color: White

Formula: C₆H₇BO₃S
MDL Number: MFCD01075681
CAS Number: 206551-43-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2ed15abe0781b4ac9f690c6b2a408c8096045de8&vid=2ed15abe0781b4ac9f690c6b2a408c8096045de8&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR