Ambroxol hydrochloride ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

A2360-25G A2360-5G
TCA2360-25GEA 499.12 CAD
TCA2360-25G TCA2360-5G
Ambroxol hydrochloride ≥98.0% (by HPLC, total nitrogen)
Ambroxol hydrochloride

CAS Number: 23828-92-4
MDL Number: MFCD00078932
Molecular Formula: C13H18Br2N2O
Molecular Weight: 414.57
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 235


Product Categories: Bioscience, Other Pharmaceutical Ingredients
Other Product Names / Synonyms: trans-4-[(2-Amino-3,5-dibromobenzyl)amino]cyclohexanol Hydrochloride

Formula: C₁₃H₁₈Br₂N₂O
Melting Pt: 235 °C
MDL Number: MFCD00078932
CAS Number: 23828-92-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=688be0fa403d87154a1c75570aa2f746537abcf2&vid=688be0fa403d87154a1c75570aa2f746537abcf2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR