2-Amino-3-pyridinemethanol ≥96.0%

Supplier: TCI America

Synonyms: (2-Amino-pyridin-3-yl)-methanol, 3-(Hydroxymethyl)-2-pyridinamine, 2-Amino-3-pyridinemethanol, 2-Amino-3-hydroxymethylpyridine, 2-Amino-3-pyridylmethanol, 2-Amino-3-(hydroxymethyl)pyridine, 2-Aminopyridine-3-methanol

A2358-1G A2358-5G
TCA2358-1GEA 79 CAD
TCA2358-1G TCA2358-5G
2-Amino-3-pyridinemethanol ≥96.0%
2-Amino-3-pyridinemethanol

CAS Number: 23612-57-9
MDL Number: MFCD05663510
Molecular Formula: C6H8N2O
Molecular Weight: 124.14
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Boiling point (°C): 279
Melting point (°C): 67


Other Product Names / Synonyms: 2-Amino-3-(hydroxymethyl)pyridine

Formula: C₆H₈N₂O
Boiling Pt: 279 °C
Melting Pt: 67 °C
MDL Number: MFCD05663510
CAS Number: 23612-57-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=073110785712faffbafce771b7b9ba1e667b08b9&vid=073110785712faffbafce771b7b9ba1e667b08b9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR