5-Amino-1,2,3-thiadiazole ≥98.0% (by HPLC)
Supplier: TCI America
A2354-25G
A2354-5G
TCA2354-25GEA
274.92
CAD
TCA2354-25G
TCA2354-5G
5-Amino-1,2,3-thiadiazole ≥98.0% (by HPLC)
5-Amino-1,2,3-thiadiazole
CAS Number: 4100-41-8
MDL Number: MFCD00082810
Molecular Formula: C2H3N3S
Molecular Weight: 101.13
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 142
Formula:
C₂H₃N₃S Melting Pt: 142 °C |
MDL Number:
MFCD00082810 CAS Number: 4100-41-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...