2-Amino-5-methylthiophene-3-carbonitrile ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2-Amino-5-methyl-3-thiophenecarbonitrile, 2-Amino-3-cyano-5-methylthiophene

A2351-25G A2351-5G
TCA2351-25GEA 156.42 CAD
TCA2351-25G TCA2351-5G
2-Amino-5-methylthiophene-3-carbonitrile ≥98.0% (by GC)
2-Amino-5-methylthiophene-3-carbonitrile

CAS Number: 138564-58-6
MDL Number: MFCD01626055
Molecular Formula: C6H6N2S
Molecular Weight: 138.19
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 103


Other Product Names / Synonyms: 2-Amino-3-cyano-5-methylthiophene

Formula: C₆H₆N₂S
MW: 138.1902 g/mol
Melting Pt: 103 °C
MDL Number: MFCD01626055
CAS Number: 138564-58-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9413b378fd5462e85d9840dd20a28e43bce46ff9&vid=9413b378fd5462e85d9840dd20a28e43bce46ff9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR