4-Amino-2-methylbenzoic acid ≥98.0%
Supplier: TCI America
Synonyms:
4-Amino-o-toluic acid, 4-Amino-2-methylbenzoic Acid
A2349-25G
A2349-5G
TCA2349-25GEA
191.18
CAD
TCA2349-25G
TCA2349-5G
4-Amino-2-methylbenzoic acid ≥98.0%
4-Amino-2-methylbenzoic acid
CAS Number: 2486-75-1
MDL Number: MFCD06656133
Molecular Formula: C8H9NO2
Molecular Weight: 151.17
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 164
Other Product Names / Synonyms: 4-Amino-o-toluic Acid
Formula:
C₈H₉NO₂ Melting Pt: 164 °C |
MDL Number:
MFCD06656133 CAS Number: 2486-75-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...