N-(4-Aminobenzoyl)-β-alanine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

A2347-25G A2347-5G
TCA2347-25GEA 90.06 CAD
TCA2347-25G TCA2347-5G
N-(4-Aminobenzoyl)-β-alanine ≥98.0% (by HPLC, titration analysis)
N-(4-Aminobenzoyl)-beta-alanine

CAS Number: 7377-08-4
MDL Number: MFCD00009805
Molecular Formula: C10H12N2O3
Molecular Weight: 208.22
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 155


Other Product Names / Synonyms: 3-(4-Aminobenzamido)propionic Acid

Formula: C₁₀H₁₂N₂O₃
Melting Pt: 155 °C
MDL Number: MFCD00009805
CAS Number: 7377-08-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2514668182a1fd7fd9fd6c487081fbf166eb197&vid=f2514668182a1fd7fd9fd6c487081fbf166eb197&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR