2-Amino-4-bromopyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

A2339-1G A2339-5G
TCA2339-1GEA 39.5 CAD
TCA2339-1G TCA2339-5G
2-Amino-4-bromopyridine ≥98.0% (by GC, titration analysis)
2-Amino-4-bromopyridine

CAS Number: 84249-14-9
MDL Number: MFCD01646115
Molecular Formula: C5H5BrN2
Molecular Weight: 173.01
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 147

Formula: C₅H₅BrN₂
Melting Pt: 147 °C
MDL Number: MFCD01646115
CAS Number: 84249-14-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=454a643a00dd92f00156c69b6a7e18143d87a3fd&vid=454a643a00dd92f00156c69b6a7e18143d87a3fd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR