2-Acetyl-4-methylpyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

A2338-1G A2338-5G
TCA2338-1GEA 137.46 CAD
TCA2338-1G TCA2338-5G
2-Acetyl-4-methylpyridine ≥98.0% (by GC, titration analysis)
2-Acetyl-4-methylpyridine

CAS Number: 59576-26-0
MDL Number: MFCD01863638
Molecular Formula: C8H9NO
Molecular Weight: 135.17
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 32
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 2-Acetyl-4-picoline

Formula: C₈H₉NO
Melting Pt: 32 °C
MDL Number: MFCD01863638
CAS Number: 59576-26-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2e9f14a2e195486efe5c50a7d8cb4eae057f806f&vid=2e9f14a2e195486efe5c50a7d8cb4eae057f806f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR