2,1-Benzisoxazole ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Anthranil, Benzo[c]isoxazole, 2,1-Benzisoxazole

A2337-1G A2337-5G
TCA2337-1GEA 58.59 CAD
TCA2337-1G TCA2337-5G
2,1-Benzisoxazole ≥98.0% (by GC)
2,1-Benzisoxazole

CAS Number: 271-58-9
MDL Number: MFCD00005806
Molecular Formula: C7H5NO
Molecular Weight: 119.12
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 101
Flash Point (°C): 94
Specific Gravity (20/20): 1.19


Other Product Names / Synonyms: 2,1-Benzisoxazole

Formula: C₇H₅NO
MW: 119.1207 g/mol
Boiling Pt: 101 °C
Density: 1.19 (20 °C)
Flash Pt: 94 °C
MDL Number: MFCD00005806
CAS Number: 271-58-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6640970e3e30926227cb803a72dc80c7d30e062&vid=f6640970e3e30926227cb803a72dc80c7d30e062&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR