2-Amino-6-methyl-3-nitropyridine ≥98.0% (by GC)

Supplier: TCI America

A2334-1G A2334-5G
TCA2334-1GEA 41.08 CAD
TCA2334-1G TCA2334-5G
2-Amino-6-methyl-3-nitropyridine ≥98.0% (by GC)
2-Amino-6-methyl-3-nitropyridine

CAS Number: 21901-29-1
MDL Number: MFCD00047443
Molecular Formula: C6H7N3O2
Molecular Weight: 153.14
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 156


Other Product Names / Synonyms: 6-Amino-5-nitro-2-picoline

Formula: C₆H₇N₃O₂
Melting Pt: 156 °C
MDL Number: MFCD00047443
CAS Number: 21901-29-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c33fd2471b26f84c366b006e69360262206fe88e&vid=c33fd2471b26f84c366b006e69360262206fe88e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR