4-Hydroxy-3-methoxybenzylamine hydrochloride ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

A2330-25G A2330-5G
TCA2330-25GEA 159.17 CAD
TCA2330-25G TCA2330-5G
4-Hydroxy-3-methoxybenzylamine hydrochloride ≥98.0% (by HPLC, total nitrogen)
4-Hydroxy-3-methoxybenzylamine hydrochloride

CAS Number: 7149-10-2
MDL Number: MFCD00012864
Molecular Formula: C8H11NO2
Molecular Weight: 189.64
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 220


Other Product Names / Synonyms: 4-Hydroxy-3-methoxybenzylamine Hydrochloride, Vanillylamine Hydrochloride

Formula: C₈H₁₂ClNO₂
Melting Pt: 220 °C
MDL Number: MFCD00012864
CAS Number: 7149-10-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7312f61d254cab2609322c2416dc8664cbf73d9d&vid=7312f61d254cab2609322c2416dc8664cbf73d9d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR