9-Anthraceneboronic acid ≥98.0% (by HPLC) (contains varying amounts of Anhydride)

Supplier: TCI America

Synonyms: 9-anthraceneboronic acid, Anthracene-9-boronic acid, anthracen-9-ylboronic acid

A2328-1G A2328-5G
TCA2328-1GEA 67.94 CAD
TCA2328-1G TCA2328-5G
9-Anthraceneboronic acid ≥98.0% (by HPLC) (contains varying amounts of Anhydride)
9-Anthraceneboronic acid

CAS Number: 100622-34-2
MDL Number: MFCD03425925
Molecular Formula: C14H11BO2
Molecular Weight: 222.05
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 217

Formula: C₁₄H₁₁BO₂
MW: 222.05 g/mol
Melting Pt: 217 °C
MDL Number: MFCD03425925
CAS Number: 100622-34-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1d46157abb45ef92684dd41a2ef65d958c5ccdb4&vid=1d46157abb45ef92684dd41a2ef65d958c5ccdb4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR