2-(2-Aminophenyl)ethanol ≥95.0% (by GC, titration analysis)

Supplier: TCI America

A2327-25G A2327-5G
TCA2327-25GEA 224.83 CAD
TCA2327-25G TCA2327-5G
2-(2-Aminophenyl)ethanol ≥95.0% (by GC, titration analysis)
2-(2-Aminophenyl)ethanol

CAS Number: 5339-85-5
MDL Number: MFCD00007754
Molecular Formula: C8H11NO
Molecular Weight: 137.18
Purity/Analysis Method: >95.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 148
Flash Point (°C): 113
Specific Gravity (20/20): 1.12


Other Product Names / Synonyms: 2-(2-Aminophenyl)ethyl Alcohol, 2-Aminophenethyl Alcohol

Formula: C₈H₁₁NO
Boiling Pt: 148 °C
Density: 1.12 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00007754
CAS Number: 5339-85-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e5fef110ca8b46a0c9532d9f8eaede75f26e5e5a&vid=e5fef110ca8b46a0c9532d9f8eaede75f26e5e5a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR