2-Amino-5-nitro-6-picoline ≥98.0%

Supplier: TCI America

Synonyms: 6-Amino-2-methyl-3-nitropyridine, 2-Amino-5-nitro-6-methylpyridine, 2-Amino-6-methyl-5-nitropyridine, 6-Amino-3-nitro-2-picoline, 6-Methyl-5-nitro-2-pyridylamine

A2326-1G A2326-5G
TCA2326-1GEA 104.12 CAD
TCA2326-1G TCA2326-5G
2-Amino-5-nitro-6-picoline ≥98.0%
2-Amino-5-nitro-6-picoline

CAS Number: 22280-62-2
MDL Number: MFCD00053582
Molecular Formula: C6H7N3O2
Molecular Weight: 153.14
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 191


Other Product Names / Synonyms: 6-Amino-3-nitro-2-picoline

Formula: C₆H₇N₃O₂
MW: 153.14
Melting Pt: 191 °C
MDL Number: MFCD00053582
CAS Number: 22280-62-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7b2e280e10fa4cbe9b74819f6e1404070d1615b6&vid=7b2e280e10fa4cbe9b74819f6e1404070d1615b6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR