(S)-3-Amino-1-carbobenzoxypyrrolidine hydrochloride ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
Synonyms:
(S)-3-Amino-1-Cbz-pyrrolidine Hydrochloride, (S)-1-Cbz-3-Aminopyrrolidine hydrochloride
CAS Number: 550378-39-7
MDL Number: MFCD06796559
Molecular Formula: C12H16N2O2
Molecular Weight: 256.73
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Flash Point (°C): 110
Specific rotation [a]20/D: 15 deg (C=1, H2O)
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (S)-3-Amino-1-Cbz-pyrrolidine Hydrochloride
Formula:
C₁₂H₁₆N₂O₂ MW: 256.73 g/mol Flash Pt: 110 °C |
MDL Number:
MFCD06796559 CAS Number: 550378-39-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...