(R)-3-Amino-1-N-Cbz-pyrrolidine hydrochloride ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
Synonyms:
(R)-3-Amino-1-carbobenzoxypyrrolidine hydrochloride, (R)-1-Cbz-3-aminopyrrolidine Hydrochloride, (R)-3-Amino-1-benzyloxycarbonylpyrrolidine hydrochloride, (R)-3-Amino-1-Cbz-pyrrolidine hydrochloride
CAS Number: 870621-17-3
MDL Number: MFCD09038802
Molecular Formula: C12H16N2O2
Molecular Weight: 256.73
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Flash Point (°C): 110
Specific rotation [a]20/D: -14 deg (C=1, H2O)
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (R)-3-Amino-1-Cbz-pyrrolidine Hydrochloride
Formula:
C₁₂H₁₇ClN₂O₂ MW: 256.7286 g/mol Flash Pt: 110 °C |
MDL Number:
MFCD09038802 CAS Number: 870621-17-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...