2-(1-Adamantyl)-4-bromoanisole ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 1-(5-Bromo-2-methoxyphenyl)adamantane, 1-(5-Bromo-2-methoxy-phenyl)adamantane

A2321-25G A2321-5G
TCA2321-25GEA 760.3 CAD
TCA2321-25G TCA2321-5G
2-(1-Adamantyl)-4-bromoanisole ≥98.0% (by GC)
2-(1-Adamantyl)-4-bromoanisole

CAS Number: 104224-63-7
MDL Number: MFCD03855308
Molecular Formula: C17H21BrO
Molecular Weight: 321.26
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 144


Other Product Names / Synonyms: 1-(5-Bromo-2-methoxyphenyl)adamantane

Formula: C₁₇H₂₁BrO
MW: 321.2520 g/mol
Melting Pt: 144 °C
MDL Number: MFCD03855308
CAS Number: 104224-63-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e74b16d12cbb86c0ae84b51e43d49fc1184a64a7&vid=e74b16d12cbb86c0ae84b51e43d49fc1184a64a7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR