3-Amino-1-Boc-piperidine ≥97.0%

Supplier: TCI America

Synonyms: 1-BOC-3-Aminopiperidine, (±)-3-Amino-1-boc-piperidine, tert-Butyl 3-aminopiperidine-1-carboxylate

A2314-1G A2314-5G
TCA2314-1GEA 44.24 CAD
TCA2314-1G TCA2314-5G
3-Amino-1-Boc-piperidine ≥97.0%
3-Amino-1-Boc-piperidine

CAS Number: 184637-48-7
MDL Number: MFCD01861219
Molecular Formula: C10H20N2O2
Molecular Weight: 200.28
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Specific Gravity (20/20): 1.02


Other Product Names / Synonyms: 3-Amino-1-Boc-piperidine, tert-Butyl 3-Amino-1-piperidinecarboxylate, 3-Amino-1-piperidinecarboxylic Acid tert-Butyl Ester

Formula: C₁₀H₂₀N₂O₂
MW: 200.28 g/mol
Density: 1.02 (20 °C)
MDL Number: MFCD01861219
CAS Number: 184637-48-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e69ba73997d4a075cb7556979f4d14a29b12c69c&vid=e69ba73997d4a075cb7556979f4d14a29b12c69c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR