N-(5-Amino-2-methylphenyl)-4-(3-pyridyl)-2-pyrimidineamine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

A2308-1G A2308-5G
TCA2308-1GEA 172.22 CAD
TCA2308-1G TCA2308-5G
N-(5-Amino-2-methylphenyl)-4-(3-pyridyl)-2-pyrimidineamine ≥98.0% (by HPLC, titration analysis)
N-(5-Amino-2-methylphenyl)-4-(3-pyridyl)-2-pyrimidineamine

CAS Number: 152460-10-1
MDL Number: MFCD09028125
Molecular Formula: C16H15N5
Molecular Weight: 277.33
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 145


Other Product Names / Synonyms: N-(5-Amino-o-tolyl)-4-(3-pyridyl)-2-pyrimidinamine, 4-Methyl-N3-[4-(3-pyridyl)-2-pyrimidinyl]-1,3-phenylenediamine

Formula: C₁₆H₁₅N₅
Melting Pt: 145 °C
MDL Number: MFCD09028125
CAS Number: 152460-10-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ca8cafbf61182710d548336fbd4ff7a8e73c21fc&vid=ca8cafbf61182710d548336fbd4ff7a8e73c21fc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR