(1S,2S)-(+)-1-Amino-2-indanol ≥98.0% (by GC)

Supplier: TCI America

A2307-1G A2307-5G
TCA2307-1GEA 110.6 CAD
TCA2307-1G TCA2307-5G
(1S,2S)-(+)-1-Amino-2-indanol ≥98.0% (by GC)
(1S,2S)-(+)-1-Amino-2-indanol

CAS Number: 163061-74-3
MDL Number: MFCD08275383
Molecular Formula: C9H11NO
Molecular Weight: 149.19
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 146
Specific rotation [a]20/D: 24 deg (C=1, MeOH)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (1S,2S)-(+)-1-Amino-2-hydroxyindan

Formula: C₉H₁₁NO
Melting Pt: 146 °C
MDL Number: MFCD08275383
CAS Number: 163061-74-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=af4a7c23fe8e93d5a422a80c0e156b84191a9420&vid=af4a7c23fe8e93d5a422a80c0e156b84191a9420&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR