(1R,2R)-(-)-1-Amino-2-indanol ≥98.0% (by GC, titration analysis)

Supplier: TCI America

A2306-1G A2306-5G
TCA2306-1GEA 139.04 CAD
TCA2306-1G TCA2306-5G
(1R,2R)-(-)-1-Amino-2-indanol ≥98.0% (by GC, titration analysis)
(1R,2R)-(-)-1-Amino-2-indanol

CAS Number: 163061-73-2
MDL Number: MFCD08275447
Molecular Formula: C9H11NO
Molecular Weight: 149.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 144
Specific rotation [a]20/D: -24 deg (C=1, MeOH)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (1R,2R)-(-)-1-Amino-2-hydroxyindan

Formula: C₉H₁₁NO
Melting Pt: 144 °C
MDL Number: MFCD08275447
CAS Number: 163061-73-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df0ce791701c3c99496cc42f775cb79c06ad2885&vid=df0ce791701c3c99496cc42f775cb79c06ad2885&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR