Allyl phenyl carbonate ≥97.0% (by GC)

Supplier: TCI America

A2303-25G A2303-5G
TCA2303-25GEA 342.86 CAD
TCA2303-25G TCA2303-5G
Allyl phenyl carbonate ≥97.0% (by GC)
Allyl phenyl carbonate

CAS Number: 16308-68-2
MDL Number: MFCD07784342
Molecular Formula: C10H10O3
Molecular Weight: 178.19
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 78
Flash Point (°C): 110
Specific Gravity (20/20): 1.11


Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis), Materials Chemistry, Monomers
Other Product Names / Synonyms: Carbonic Acid Allyl Phenyl Ester

Formula: C₁₀H₁₀O₃
Boiling Pt: 78 °C
Density: 1.11 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD07784342
CAS Number: 16308-68-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=55474e206497ac68bddb533c6b74efbf57caeb9b&vid=55474e206497ac68bddb533c6b74efbf57caeb9b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR