2-Amino-5-fluorophenol ≥97.0% (by GC, titration analysis)

Supplier: TCI America

A2300-1G A2300-5G
TCA2300-1GEA 61.62 CAD
TCA2300-1G TCA2300-5G
2-Amino-5-fluorophenol ≥97.0% (by GC, titration analysis)
2-Amino-5-fluorophenol

CAS Number: 53981-24-1
MDL Number: MFCD00671759
Molecular Formula: C6H6FNO
Molecular Weight: 127.12
Purity/Analysis Method: >97.0% (GC)
Form: Crystal


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-Fluoro-2-hydroxyaniline

Formula: C₆H₆FNO
MDL Number: MFCD00671759
CAS Number: 53981-24-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b807a96d296878fe7b03e999f54b23f4b7852c3f&vid=b807a96d296878fe7b03e999f54b23f4b7852c3f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR