4-Acetylindan ≥98.0%

Supplier: TCI America

Synonyms: 1-(2,3-Dihydro-1H-inden-4-yl)ethanone, 1-Indan-4-yl-ethanone

A2296-1G A2296-5G
TCA2296-1GEA 158 CAD
TCA2296-1G TCA2296-5G
4-Acetylindan ≥98.0%
4-Acetylindan

CAS Number: 38997-97-6
MDL Number: MFCD09033715
Molecular Formula: C11H12O
Molecular Weight: 160.22
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 103
Melting point (°C): 41

Formula: C₁₁H₁₂O
Boiling Pt: 103 °C
Melting Pt: 41 °C
MDL Number: MFCD09033715
CAS Number: 38997-97-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6c0550927594a3d618516e3e95f2ad33417eed5&vid=f6c0550927594a3d618516e3e95f2ad33417eed5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR