4-Aminocyclohexaneethanol (cis- and trans- mixture) 98,0

Supplier: TCI America

Synonyms: 1-Amino-4-(2-hydroxyethyl)cyclohexane, 4-Aminocyclohexaneethanol, 2-(4-Aminocyclohexyl)ethanol

A2288-1G A2288-5G
TCA2288-1GEA 224.91 CAD
TCA2288-1G TCA2288-5G
4-Aminocyclohexaneethanol (cis- and trans- mixture) 98,0
4-Aminocyclohexaneethanol (cis- and trans- mixture)

CAS Number: 857831-26-6
Molecular Formula: C8H17NO
Molecular Weight: 143.23
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 85


Other Product Names / Synonyms: 2-(4-Aminocyclohexyl)ethanol, 1-Amino-4-(2-hydroxyethyl)cyclohexane

Formula: C₈H₁₇NO
MW: 143.23 g/mol
Melting Pt: 85 °C
MDL Number: MFCD15072144
CAS Number: 857831-26-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=767b43ed3ba2b59d7502a75f0eecd80d62d564ee&vid=767b43ed3ba2b59d7502a75f0eecd80d62d564ee&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR